1,3-Difluoro-2-iodo-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

51222-1G 51222-500MG 51222-5G
76891-000EA 560.16 USD
76891-000 76891-002 76891-004
1,3-Difluoro-2-iodo-5-methoxybenzene ≥95%
1,3-Difluoro-2-iodo-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
Storage Temperature: Ambient
MDL Number: MFCD20040388
CAS Number: 1140622-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR