(2-Chloro-6-methoxyphenyl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

50700-1G 50700-25G 50700-500MG 50700-5G
76890-958EA 206.88 USD
76890-958 76890-960 76890-962 76890-964
(2-Chloro-6-methoxyphenyl)methanol ≥97% research grade
(2-Chloro-6-methoxyphenyl)methanol
Formula: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11190334
CAS Number: 1038726-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR