3-Bromo-2,6-difluorobenzal fluoride ≥97%

Supplier: AOBChem USA

Warning

50597-1G 50597-25G 50597-500MG 50597-5G
76890-936EA 126.25 USD
76890-936 76890-938 76890-940 76890-942
3-Bromo-2,6-difluorobenzal fluoride ≥97%
3-Bromo-2,6-difluorobenzal fluoride
Formula: C₇H₃BrF₄
Storage Temperature: Refrigerator
MDL Number: MFCD25976517
CAS Number: 1447671-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR