5-Bromo-2,4-dichlorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-2,4-dichlorophenyl)methanol

50161-1G 50161-25G 50161-500MG 50161-5G
76890-924EA 274.78 USD
76890-924 76890-926 76890-928 76890-930
5-Bromo-2,4-dichlorobenzyl alcohol ≥97%
5-Bromo-2,4-dichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MW: 255.924 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28737839
CAS Number: 1805023-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR