1-Methyl-1H-pyrazole-5-carboxamide ≥97%
Supplier: AOBChem USA
|
Warning
|
50101-1G
50101-25G
50101-500MG
50101-5G
76890-910EA
144.29
USD
76890-910
76890-912
76890-914
76890-916
1-Methyl-1H-pyrazole-5-carboxamide ≥97%
1-Methyl-1H-pyrazole-5-carboxamide
Formula:
C₅H₇N₃O Storage Temperature: Refrigerator |
MDL Number:
MFCD00460857 CAS Number: 98711-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...