2,4-Dichloro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49937-1G 49937-500MG 49937-5G
76890-868EA 500.75 USD
76890-868 76890-870 76890-872
2,4-Dichloro-3-methylbenzaldehyde ≥97%
2,4-Dichloro-3-methylbenzaldehyde
Formula: C₈H₆Cl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD24679545
CAS Number: 63525-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR