(3,5-Dibromophenyl)(phenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

49877-1G 49877-25G 49877-500MG 49877-5G
76890-852EA 233.4 USD
76890-852 76890-854 76890-856 76890-858
(3,5-Dibromophenyl)(phenyl)methanone ≥97%
(3,5-Dibromophenyl)(phenyl)methanone
Formula: C₁₃H₈Br₂O
MW: 340.01 g/mol
Storage Temperature: Ambient
CAS Number: 51339-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR