(2,6-Dibromophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49709-25G 49709-5G
76890-846EA 563.34 USD
76890-846 76890-848
(2,6-Dibromophenyl)methanol ≥97%
(2,6-Dibromophenyl)methanol
Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185492
CAS Number: 1013031-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR