2-(2-Fluoro-5-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

49617-25G 49617-5G
76890-816EA 257.8 USD
76890-816 76890-818
2-(2-Fluoro-5-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(2-Fluoro-5-(trifluoromethyl)phenyl)-1,3-dioxolane
Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209161
CAS Number: 773102-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR