1-Bromo-3-((4-fluorobenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

49580-1G 49580-25G 49580-5G
76890-780EA 62.6 USD
76890-780 76890-782 76890-784
1-Bromo-3-((4-fluorobenzyl)oxy)benzene ≥97%
1-Bromo-3-((4-fluorobenzyl)oxy)benzene
Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Storage Temperature: Ambient
CAS Number: 845866-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR