2,4-Dimethoxy-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

49453-25G 49453-5G
76890-758EA 346.92 USD
76890-758 76890-760
2,4-Dimethoxy-1-fluorobenzene ≥97%
2,4-Dimethoxy-1-fluorobenzene
Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD00042265
CAS Number: 17715-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR