2-(2-Fluoro-5-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

49349-25G 49349-5G
76890-732EA 360.71 USD
76890-732 76890-734
2-(2-Fluoro-5-methylphenyl)-1,3-dioxolane ≥97%
2-(2-Fluoro-5-methylphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209624
CAS Number: 773095-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR