2,3-Difluoro-4-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49344-25G 49344-5G
76890-720EA 608.96 USD
76890-720 76890-722
2,3-Difluoro-4-hydroxybenzaldehyde ≥97%
2,3-Difluoro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08235194
CAS Number: 676500-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR