2-Bromo-4-chloro-5-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-5-chloro-2-methyl-4-nitrobenzene

49197-1G 49197-25G 49197-500MG 49197-5G
76890-698EA 347.98 USD
76890-698 76890-700 76890-702 76890-704
2-Bromo-4-chloro-5-nitrotoluene ≥97%
2-Bromo-4-chloro-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00051512
CAS Number: 40371-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR