2-Bromo-4-fluoro-5-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-5-fluoro-2-methyl-4-nitrobenzene

49193-25G 49193-5G
76890-686EA 346.92 USD
76890-686 76890-688
2-Bromo-4-fluoro-5-nitrotoluene ≥97%
2-Bromo-4-fluoro-5-nitrotoluene
Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847636
CAS Number: 64695-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR