1-(2,4,6-Trifluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

49154-1G 49154-25G 49154-500MG 49154-5G
76890-676EA 144.29 USD
76890-676 76890-678 76890-680 76890-682
1-(2,4,6-Trifluorophenyl)ethanol ≥97%
1-(2,4,6-Trifluorophenyl)ethanol
Formula: C₈H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD12787167
CAS Number: 1250113-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR