2-(2-Fluoro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

48993-1G 48993-25G 48993-500MG 48993-5G
76890-646EA 245.07 USD
76890-646 76890-648 76890-650 76890-652
2-(2-Fluoro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Fluoro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₄H₂₀BFO₂
MW: 250.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914525
CAS Number: 2096341-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR