1-Bromo-2-fluoro-3-iodo-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

48815-1G 48815-500MG 48815-5G
76890-632EA 558.04 USD
76890-632 76890-634 76890-636
1-Bromo-2-fluoro-3-iodo-4-(trifluoromethyl)benzene ≥97%
1-Bromo-2-fluoro-3-iodo-4-(trifluoromethyl)benzene
Formula: C₇H₂BrF₄I
MW: 368.893 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29049271
CAS Number: 1935625-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR