2-(Benzyloxy)-1-bromo-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

48760-1G 48760-25G 48760-500MG 48760-5G
76890-598EA 155.96 USD
76890-598 76890-600 76890-602 76890-604
2-(Benzyloxy)-1-bromo-4-iodobenzene ≥97%
2-(Benzyloxy)-1-bromo-4-iodobenzene
Formula: C₁₃H₁₀BrIO
Storage Temperature: Ambient
MDL Number: MFCD33403904
CAS Number: 2253887-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR