1-Fluoro-4-(4-iodophenoxy)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-4'-iododiphenyl ether

48730-1G 48730-25G 48730-5G
76890-584EA 91.24 USD
76890-584 76890-586 76890-588
1-Fluoro-4-(4-iodophenoxy)benzene ≥97% research grade
1-Fluoro-4-(4-iodophenoxy)benzene
Formula: C₁₂H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD07368761
CAS Number: 886762-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR