2,3,6-Trichloroiodobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1,2,4-trichloro-3-iodobenzene

48677-25G 48677-5G
76890-576EA 360.71 USD
76890-576 76890-578
2,3,6-Trichloroiodobenzene ≥97% research grade
2,3,6-Trichloroiodobenzene
Formula: C₆H₂Cl₃I
MW: 307.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13176521
CAS Number: 38411-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR