1-Bromo-2,5-dichloro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

48659-1G 48659-25G 48659-500MG 48659-5G
76890-564EA 124.13 USD
76890-564 76890-566 76890-568 76890-570
1-Bromo-2,5-dichloro-4-methoxybenzene ≥97%
1-Bromo-2,5-dichloro-4-methoxybenzene
Formula: C₇H₅BrCl₂O
MW: 255.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21993562
CAS Number: 55026-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR