2,4,6-trifluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48650-1G 48650-500MG 48650-5G
76890-550EA 525.15 USD
76890-550 76890-552 76890-554
2,4,6-trifluoro-3-hydroxybenzaldehyde ≥97%
2,4,6-trifluoro-3-hydroxybenzaldehyde
Formula: C₇H₃F₃O₂
MW: 176.093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24686494
CAS Number: 1093253-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR