1-Bromo-2,3-difluoro-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

48601-1G 48601-25G 48601-5G
76890-540EA 102.91 USD
76890-540 76890-542 76890-544
1-Bromo-2,3-difluoro-4-methylbenzene ≥95%
1-Bromo-2,3-difluoro-4-methylbenzene
Formula: C₇H₅BrF₂
MW: 207.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21603711
CAS Number: 928304-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR