2'-Hydroxy-5'-methyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,2,2-trifluoro-1-(2-hydroxy-5-methylphenyl)ethanone

48467-1G 48467-500MG 48467-5G
76890-512EA 1163.81 USD
76890-512 76890-514 76890-516
2'-Hydroxy-5'-methyl-2,2,2-trifluoroacetophenone ≥97%
2'-Hydroxy-5'-methyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD05864383
CAS Number: 70978-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR