1,3-Dibromo-5-(tert-butyl)-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

48443-1G 48443-25G 48443-500MG 48443-5G
76890-498EA 165.51 USD
76890-498 76890-500 76890-502 76890-504
1,3-Dibromo-5-(tert-butyl)-2-iodobenzene ≥95%
1,3-Dibromo-5-(tert-butyl)-2-iodobenzene
Formula: C₁₀H₁₁Br₂I
Storage Temperature: Ambient
MDL Number: MFCD09878147
CAS Number: 172368-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR