1,2,3-Tribromo-5-(tert-butyl)benzene ≥95% research grade

Supplier: AOBChem USA

Warning

48441-1G 48441-25G 48441-5G
76890-484EA 62.6 USD
76890-484 76890-486 76890-488
1,2,3-Tribromo-5-(tert-butyl)benzene ≥95% research grade
1,2,3-Tribromo-5-(tert-butyl)benzene
Formula: C₁₀H₁₁Br₃
Storage Temperature: Ambient
MDL Number: MFCD33403885
CAS Number: 114895-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR