(4-Methoxy-3,5-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

48363-1G 48363-25G 48363-500MG 48363-5G
76890-468EA 233.4 USD
76890-468 76890-470 76890-472 76890-474
(4-Methoxy-3,5-dimethylphenyl)methanol ≥97%
(4-Methoxy-3,5-dimethylphenyl)methanol
Formula: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201142
CAS Number: 83037-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR