1-Fluoro-2-(methoxymethoxy)-3-methylbenzene 95%
Supplier: AOBChem USA
|
Warning
|
48319-1G
48319-25G
48319-5G
48319-500MG
76890-454EA
114.58
USD
76890-454
76890-456
76890-458
77651-256
1-Fluoro-2-(methoxymethoxy)-3-methylbenzene 95%
1-Fluoro-2-(methoxymethoxy)-3-methylbenzene
Precaución: for Research Use Only
Formula:
C₉H₁₁FO₂ MW: 170.181 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD34756428 CAS Number: 2807469-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...