4-Bromo-2,3-dichloroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2,3-dichloro-4-iodobenzene

48293-25G 48293-5G
76890-444EA 630.18 USD
76890-444 76890-446
4-Bromo-2,3-dichloroiodobenzene ≥97%
4-Bromo-2,3-dichloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂I
Storage Temperature: Ambient
CAS Number: 1000574-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR