3-Bromo-4-fluorothioanisole ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-4-fluorophenyl)(methyl)sulphane

48265-1G 48265-25G 48265-500MG 48265-5G
76890-430EA 241.89 USD
76890-430 76890-432 76890-434 76890-436
3-Bromo-4-fluorothioanisole ≥95% research grade
3-Bromo-4-fluorothioanisole
Formula: C₇H₆BrFS
Storage Temperature: Ambient
MDL Number: MFCD22488902
CAS Number: 1511032-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR