(4-Cyclopentyl-3-fluorophenyl)boronic acid 97% research grade

Supplier: AOBChem USA

Warning

48111-1G 48111-500MG 48111-5G 48111-25G
76890-408EA 825.39 USD
76890-408 76890-410 76890-412 77656-112
(4-Cyclopentyl-3-fluorophenyl)boronic acid 97% research grade
(4-Cyclopentyl-3-fluorophenyl)boronic acid
Formula: C₁₁H₁₄BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32707296
CAS Number: 2225178-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR