1,2,5-Trichloro-3-nitrobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

47773-25G 47773-5G
76890-366EA 328.88 USD
76890-366 76890-368
1,2,5-Trichloro-3-nitrobenzene ≥97% research grade
1,2,5-Trichloro-3-nitrobenzene
Formula: C₆H₂Cl₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00456797
CAS Number: 34283-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR