2-(2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

47754-1G 47754-25G 47754-500MG 47754-5G
76890-344EA 371.32 USD
76890-344 76890-346 76890-348 76890-350
2-(2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propan-2-ol ≥95%
2-(2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propan-2-ol
Formula: C₁₅H₂₂BClO₃
MW: 296.597 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18730081
CAS Number: 885069-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR