(2-Chloro-3-methoxypyridin-4-yl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-3-methoxypyridine-4-boronic acid

47537-1G 47537-25G 47537-500MG 47537-5G
76890-318EA 231.28 USD
76890-318 76890-320 76890-322 76890-324
(2-Chloro-3-methoxypyridin-4-yl)boronic acid ≥95% research grade
(2-Chloro-3-methoxypyridin-4-yl)boronic acid
Formula: C₆H₇BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD16610616
CAS Number: 2096339-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR