3-Ethoxy-2,6-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

47522-1G 47522-25G 47522-5G
76890-292EA 64.72 USD
76890-292 76890-294 76890-296
3-Ethoxy-2,6-difluorobenzyl alcohol ≥97%
3-Ethoxy-2,6-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11519370
CAS Number: 1092461-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR