(5-Bromo-2-chloropyridin-3-yl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

47428-1G 47428-25G 47428-500MG 47428-5G
76890-280EA 251.44 USD
76890-280 76890-282 76890-284 76890-286
(5-Bromo-2-chloropyridin-3-yl)(4-methylpiperazin-1-yl)methanone ≥97%
(5-Bromo-2-chloropyridin-3-yl)(4-methylpiperazin-1-yl)methanone
Formula: C₁₁H₁₃BrClN₃O
MW: 318.597 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15504362
CAS Number: 1457225-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR