1-Bromo-2-(cyclohexylmethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

47236-1G 47236-500MG 47236-5G
76890-258EA 558.04 USD
76890-258 76890-260 76890-262
1-Bromo-2-(cyclohexylmethyl)benzene ≥97%
1-Bromo-2-(cyclohexylmethyl)benzene
Formula: C₁₃H₁₇Br
MW: 253.178 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707249
CAS Number: 1781176-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR