1-Bromo-4-(phenoxymethyl)benzene 95%

Supplier: AOBChem USA

Warning

46925-1G 46925-25G 46925-5G 46925-500MG
76890-248EA 75.33 USD
76890-248 76890-250 76890-252 77647-078
1-Bromo-4-(phenoxymethyl)benzene 95%
1-Bromo-4-(phenoxymethyl)benzene
Formula: C₁₃H₁₁BrO
MW: 263.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017847
CAS Number: 20600-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR