2-(2-Fluoro-5-iodophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

46891-25G 46891-5G
76890-240EA 240.83 USD
76890-240 76890-242
2-(2-Fluoro-5-iodophenyl)-1,3-dioxolane ≥97%
2-(2-Fluoro-5-iodophenyl)-1,3-dioxolane
Formula: C₉H₈FIO₂
MW: 294.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209631
CAS Number: 773095-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR