1-Methyl-1H-pyrazole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Methylpyrazole-5-carboxaldehyde, 5-Formyl-1-methylpyrazole

46678-25G 46678-5G
76890-216EA 198.39 USD
76890-216 76890-218
1-Methyl-1H-pyrazole-5-carbaldehyde ≥97%
1-Methyl-1H-pyrazole-5-carbaldehyde
Formula: C₅H₆N₂O
MW: 110.1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419801
CAS Number: 27258-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR