(5-Bromo-4-fluoro-2-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

46320-1G 46320-25G 46320-500MG 46320-5G
76890-160EA 257.8 USD
76890-160 76890-162 76890-164 76890-166
(5-Bromo-4-fluoro-2-methoxyphenyl)methanol ≥97%
(5-Bromo-4-fluoro-2-methoxyphenyl)methanol
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23701734
CAS Number: 923281-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR