1-Bromo-4-chloro-5-fluoro-2-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46011-1G 46011-500MG 46011-5G 46011-25G
76890-078EA 495.45 USD
76890-078 76890-080 76890-082 77655-870
1-Bromo-4-chloro-5-fluoro-2-(trifluoromethyl)benzene ≥95%
1-Bromo-4-chloro-5-fluoro-2-(trifluoromethyl)benzene
Formula: C₇H₂BrClF₄
MW: 277.441 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31558281
CAS Number: 2169320-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR