(3-Bromo-5-fluorophenyl)(piperidin-1-yl)methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
45942-1G
45942-25G
45942-500MG
45942-5G
76890-048EA
144.29
USD
76890-048
76890-050
76890-052
76890-054
(3-Bromo-5-fluorophenyl)(piperidin-1-yl)methanone ≥95%
(3-Bromo-5-fluorophenyl)(piperidin-1-yl)methanone
Formula:
C₁₂H₁₃BrFNO MW: 286.1401 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20541857 CAS Number: 1326982-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...