2-(2,4-Difluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

45930-1G 45930-25G 45930-500MG 45930-5G
76890-034EA 153.84 USD
76890-034 76890-036 76890-038 76890-040
2-(2,4-Difluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,4-Difluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₇BF₂O₃
MW: 270.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28061287
CAS Number: 1809298-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR