2-(2-Chloro-6-fluoro-3-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

45904-25G 45904-5G
76890-022EA 453.01 USD
76890-022 76890-024
2-(2-Chloro-6-fluoro-3-methylphenyl)-1,3-dioxolane ≥95%
2-(2-Chloro-6-fluoro-3-methylphenyl)-1,3-dioxolane
Formula: C₁₀H₁₀ClFO₂
MW: 216.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209310
CAS Number: 773093-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR