2-(4-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

45624-1G 45624-25G 45624-5G 45624-500MG
76889-994EA 114.58 USD
76889-994 76889-996 76889-998 77650-902
2-(4-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(4-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₉H₂₁BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD32902679
CAS Number: 2246646-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR