2-(4-Chlorophenyl)-2-oxoethyl acetate ≥97%

Supplier: AOBChem USA

Warning

45583-1G 45583-25G 45583-500MG 45583-5G
76889-978EA 147.47 USD
76889-978 76889-980 76889-982 76889-984
2-(4-Chlorophenyl)-2-oxoethyl acetate ≥97%
2-(4-Chlorophenyl)-2-oxoethyl acetate
Formula: C₁₀H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD00662320
CAS Number: 39561-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR