(3-Ethoxy-4-fluorophenyl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

45533-1G 45533-25G 45533-500MG 45533-5G
76889-968EA 343.74 USD
76889-968 76889-970 76889-972 76889-974
(3-Ethoxy-4-fluorophenyl)methanol ≥97% research grade
(3-Ethoxy-4-fluorophenyl)methanol
Formula: C₉H₁₁FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19440324
CAS Number: 1000207-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR