1,5-Dibromo-2,3-dimethylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
45423-1G
45423-25G
45423-500MG
45423-5G
76889-946EA
135.8
USD
76889-946
76889-948
76889-950
76889-952
1,5-Dibromo-2,3-dimethylbenzene ≥97%
1,5-Dibromo-2,3-dimethylbenzene
Formula:
C₈H₈Br₂ MW: 263.957 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09800832 CAS Number: 51209-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...